missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S1890

Catalog No. S1890


Only null left

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Ethanolamine141-43-51000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Ethanolamine
141-43-5
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
C2H7NO
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
HZAXFHJVJLSVMW-UHFFFAOYSA-N
Ethanol
61.08
700
2mL amber prescored glass ampule
0.1
0.1
MFCD00008183
Certified Reference Material
NCCO
2-aminoethanol
1 mL
CHEBI:16000

Chemical Identifiers

141-43-5
61.08
HZAXFHJVJLSVMW-UHFFFAOYSA-N
CHEBI:16000
NCCO
C2H7NO
MFCD00008183
700
2-aminoethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. Harmful in contact with skin. Causes serious eye irritation. Harmful if inhaled.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only