missing translation for 'onlineSavingsMsg'
Learn More

Ethacrynic acid

Catalog No. aaj6368403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6368403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6368403 Supplier Thermo Scientific Chemicals Supplier No. J6368403

CAS: 58-54-8 | C13H12Cl2O4 | 303.135 g/mol

Ethacrynic acid is used to inhibits symport of sodium, potassium, and chloride primarily in the ascending limb of Henle, but also in the proximal and distal tubules. This pharmacological action results in excretion of these ions, increased urinary output, and reduction in extracellular fluid. This compound has been classified as a loop or high ceiling diuretic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethacrynic acid is used to inhibits symport of sodium, potassium, and chloride primarily in the ascending limb of Henle, but also in the proximal and distal tubules. This pharmacological action results in excretion of these ions, in

Chemical Identifiers

CAS 58-54-8
Molecular Formula C13H12Cl2O4
Molecular Weight (g/mol) 303.135
MDL Number MFCD00056693
InChI Key AVOLMBLBETYQHX-UHFFFAOYSA-N
Synonym ethacrynic acid, etacrynic acid, ethacrynate, etacrinic acid, hydromedin, edecrin, crinuryl, edecril, edecrina, endecril
PubChem CID 3278
ChEBI CHEBI:4876
IUPAC Name 2-[2,3-dichloro-4-(2-methylidenebutanoyl)phenoxy]acetic acid
SMILES CCC(=C)C(=O)C1=C(C(=C(C=C1)OCC(=O)O)Cl)Cl

Specifications

Color White
Quantity 1 g
Merck Index 14,3717
Solubility Information Soluble in ethanol,chloroform,ether,ammonia,carbonates,and methanol. Insoluble in water.
Formula Weight 303.14
Physical Form Crystalline Powder
Chemical Name or Material Ethacrynic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-384-1
RTECSNumber AG6600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only