missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etamsylate 98.0+%, TCI America™
$55.92 - $165.90
Chemical Identifiers
| CAS | 2624-44-4 |
|---|---|
| Molecular Formula | C10H17NO5S |
| Molecular Weight (g/mol) | 263.308 |
| MDL Number | MFCD00867499 |
| InChI Key | HBGOLJKPSFNJSD-UHFFFAOYSA-N |
| Synonym | Diethylamine 2,5-Dihydroxybenzenesulfonate |
| PubChem CID | 17506 |
| IUPAC Name | 2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine |
| SMILES | CCNCC.C1=CC(=C(C=C1O)S(=O)(=O)O)O |
Chemical Identifiers
| 2624-44-4 | |
| 263.308 | |
| HBGOLJKPSFNJSD-UHFFFAOYSA-N | |
| 17506 | |
| CCNCC.C1=CC(=C(C=C1O)S(=O)(=O)O)O |
| C10H17NO5S | |
| MFCD00867499 | |
| Diethylamine 2,5-Dihydroxybenzenesulfonate | |
| 2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine |
Specifications
| 2624-44-4 | |
| White | |
| MFCD00867499 | |
| Diethylamine 2,5-Dihydroxybenzenesulfonate | |
| CCNCC.C1=CC(=C(C=C1O)S(=O)(=O)O)O | |
| 263.308 | |
| 263.31 | |
| Crystalline Powder |
| 130°C | |
| C10H17NO5S | |
| 1 g | |
| HBGOLJKPSFNJSD-UHFFFAOYSA-N | |
| 2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine | |
| 17506 | |
| ≥98.0% (HPLC,T) | |
| Etamsylate |
Safety and Handling
RTECSNumber : DB6145000
TSCA : Yes