missing translation for 'onlineSavingsMsg'
Learn More

Etamsylate 98.0+%, TCI America™

Numéro de catalogue. E11451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E11451G 1 g
E11455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E11451G Fournisseur TCI America Code fournisseur E11451G
Il en reste null

Identifiants chimiques

CAS 2624-44-4
Molecular Formula C10H17NO5S
Molecular Weight (g/mol) 263.308
MDL Number MFCD00867499
InChI Key HBGOLJKPSFNJSD-UHFFFAOYSA-N
Synonym Diethylamine 2,5-Dihydroxybenzenesulfonate
PubChem CID 17506
IUPAC Name 2,5-dihydroxybenzenesulfonic acid;N-ethylethanamine
SMILES CCNCC.C1=CC(=C(C=C1O)S(=O)(=O)O)O

Spécifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 263.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Etamsylate
RTECSNumber DB6145000
TSCA Yes