missing translation for 'onlineSavingsMsg'
Learn More

Medisca Estriol USP (Micronized)

Catalog No. 14186217
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186217 1 kg
14185250 5 g
14185252 25 g
14185660 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14186217 Supplier Medisca Supplier No. 073209

Estriol USP is a white to practically white, odorless crystalline powder.

Estriol USP is a white to practically white, odorless crystalline powder. It is insoluble in water, is sparingly soluble in alcohol, and is soluble in vegetable oils. Micronized Estriol USP has over 90% of particles that are less than 10 μm in size.

Alternative name(s): Oestriol

Specifications

Chemical Name or Material Estriol USP (Micronized)
Packaging Jar 3800ml, White, HDPE, Round, 110-400, TRIVENI-DB, CT
CAS 50-27-1
Type Drug Standard
Molecular Weight (g/mol) 288.38
Quantity 1 kg
Percent Purity 97 to 102 %
Grade USP