missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Estriol USP (Micronized)
Description
Estriol USP is a white to practically white, odorless crystalline powder. It is insoluble in water, is sparingly soluble in alcohol, and is soluble in vegetable oils. Micronized Estriol USP has over 90% of particles that are less than 10 μm in size.
Alternative name(s): Oestriol
Specifications
Specifications
| Chemical Name or Material | Estriol USP (Micronized) |
| Packaging | Jar 30ml, White, HDPE, Round, 28-400, TRIVENI-DB, CT |
| CAS | 50-27-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 288.38 |
| Quantity | 5 g |
| Percent Purity | 97 to 102 % |
| Grade | USP |