Learn More
Invitrogen™ Estriol ELISA Kit
Description
The Estriol Competitive ELISA quantitates estriol in serum, plasma, urine, saliva, dried fecal extracts or or cell culture medium.∣Principle of the method: The Estriol Competitive ELISA research-use-only kit is designed to quantitatively measure estriol independent of species. An estriol standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit estriol antibodies and estriol-peroxidase conjugate is added to the wells. The estriol-peroxidase conjugate and any estriol in the sample will compete to bind to the anti-rabbit estriol antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound estriol-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of estriol present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | C18H24O3; Estratriol; Estriol; Oestriol; Ovestin; Ovestrion |
| Assay Range | 19.66-12000 pg/mL |
| Assay Sensitivity | 5.33 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Fecal Extract, Plasma, Serum, Supernatant, Urine |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | C18H24O3; Estratriol; Estriol; Oestriol; Ovestin; OvestrionC18H24O3; Estratriol; Estriol; Oestriol; Ovestin; Ovestrion; |
| Show More |