missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal 98.0+%, TCI America™
$54.67 - $164.03
Chemical Identifiers
| CAS | 5571-36-8 |
|---|---|
| Molecular Formula | C20H26O3 |
| Molecular Weight (g/mol) | 314.425 |
| MDL Number | MFCD08062585 |
| InChI Key | XUOQKQRMICQUQC-AOIWGVFYSA-N |
| PubChem CID | 7076399 |
| IUPAC Name | (8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one |
| SMILES | CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5 |
Chemical Identifiers
| 5571-36-8 | |
| 314.425 | |
| XUOQKQRMICQUQC-AOIWGVFYSA-N | |
| (8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one |
| C20H26O3 | |
| MFCD08062585 | |
| 7076399 | |
| CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5 |
Specifications
| 5571-36-8 | |
| White-Yellow | |
| MFCD08062585 | |
| XUOQKQRMICQUQC-AOIWGVFYSA-N | |
| (8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one | |
| 7076399 | |
| ≥98.0% (HPLC) | |
| Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal |
| 156°C | |
| C20H26O3 | |
| 3077 | |
| CC12CC=C3C(C1CCC2=O)CCC4=C3CCC5(C4)OCCO5 | |
| 314.425 | |
| 314.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No