missing translation for 'onlineSavingsMsg'
Learn More

Esomeprazole Magnesium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013551
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013551 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013551 Supplier Merck Emd Millipore Supplier No. PHR15851G
Esomeprazole Magnesium is theS-isomer of omeprazole and is widely used for the treatment of acid related diseases. It is the first single optical isomer proton pump inhibitor and is found to be better over omeprazole in terms of pharmacokinetic characteristics and acid suppression.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 217087-09-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C34H36 mgN6O6S2 · 3H2 O
Synonym (S)-Omeprazole magnesium trihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 767.17
Grade Certified Reference Material