missing translation for 'onlineSavingsMsg'
Learn More

Esmolol 98.0+%, TCI America™

Catalog No. E1106100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1106100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1106100MG Supplier TCI America Supplier No. E1106100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity crystalline powder intended for laboratory research applications.

Chemical Identifiers

CAS 81147-92-4
Molecular Formula C16H26ClNO4
Molecular Weight (g/mol) 331.84
MDL Number MFCD00864566
InChI Key GEKNCWBANDDJJL-UHFFFAOYNA-N
Synonym Methyl 3-[4-[2-Hydroxy-3-(isopropylamino)propoxy]phenyl]propionate, 3-[4-[2-Hydroxy-3-(isopropylamino)propoxy]phenyl]propionic Acid Methyl Ester
PubChem CID 59768
ChEBI CHEBI:4856
IUPAC Name hydrogen methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate chloride
SMILES [H+].[Cl-].COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1

Specifications

Melting Point 61°C
Color White
Quantity 100 mg
Formula Weight 295.38
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Esmolol
RTECSNumber DA8345500
TSCA No