accessibility menu, dialog, popup

UserName

CAS RN 81147-92-4

IUPAC Name: hydrogen methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate chloride
Synonyms: hydrogen esmolol chloride
Molecular Weight (g/mol): 331.84
Molecular Formula: C16H26ClNO4
InChi Key: GEKNCWBANDDJJL-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results