missing translation for 'onlineSavingsMsg'
Learn More

Escitalopram Oxalate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013562
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013562 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013562 Supplier Merck Emd Millipore Supplier No. PHR17331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Escitalopram Oxalate, a highly selective serotonin re-uptake inhibitor antidepressant, is a pure S-enantiomer of the racemic, bicyclic pthalates derivatives of citalopram. It is mainly developed for the treatment of depression and anxiety disorders. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 219861-08-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H21 FN2 O · C2H2O4
MDL Number MFCD06407826
Synonym Escitalopram oxalate; S-(+)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate; S-(+)-Citalopram oxalate
Type Certified Reference Material
Quantity 1 g
Formula Weight 414.43
Grade Certified Reference Material