missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B, spirit soluble

CAS: 15905-32-5 | C20H8I4O5 | 835.90 g/mol

Supplier:  Thermo Scientific Chemicals J6261914

 View more versions of this product

Catalog No. AAJ6261914


Only null left

Description

Description

Erythrosin B is used as a red coloring in some foods such as cherries and fish. It is used as printing ink, dental plaque disclosing agent and biological stain. It finds application as a radiopaque medium and sensitizer for orthochromatic photographic films. It is utilized as a cake-decorating gel. In addition to this, it is used in sweets such as candies and popsicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B is used as a red coloring in some foods such as cherries and fish. It is used as printing ink, dental plaque disclosing agent and biological stain. It finds application as a radiopaque medium and sensitizer for orthochromatic photographic films. It is utilized as a cake-decorating gel. In addition to this, it is used in sweets such as candies and popsicles.

Solubility
Soluble in water, spirit, methanol, methyl cellosolve and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Erythrosin B
C20H8I4O5
C.I. 45430:2; Solvent Red 140
OALHHIHQOFIMEF-UHFFFAOYSA-N
3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
3259
Orange to Orange/Red
Powder
15905-32-5
MFCD00005044
Soluble in water,spirit,methanol,methyl cellosolve and ethanol.
OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2
835.90
835.89
25 g

Chemical Identifiers

15905-32-5
835.90
OALHHIHQOFIMEF-UHFFFAOYSA-N
3259
C20H8I4O5
MFCD00005044
C.I. 45430:2; Solvent Red 140
OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 240-046-0

RTECSNumber : LM5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only