missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Erythrosin B, pure, certified
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Absorption Ratio | 1.15 to 1.55 |
| Dye Content | 80% min. |
| Infrared Spectrum | Authentic |
| Lambda Max | 524 to 527nm |
| Packaging | Glass bottle |
| Beilstein | 19, II, 258 |
| Merck Index | 15, 3749 |
| Color | Brown to Red |
| Quantity | 25 g |
| Physical Form | Fine Powder |
| Show More |
Frequently Asked Questions (FAQs)
What is the alternative to TrypanBlue for using it with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000?
The suitable alternative would be Erythrosin B (Cat. No. 409451000, A14180.36, 189460250 and J62619.22).
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement: Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only