missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Erythromycin, Eur.Ph.

Catalog No. AC343880050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC343880050 5 g Glass Bottle
AC343880250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC343880050 Supplier Thermo Scientific Chemicals Supplier No. 343880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Erythromycin (E-Mycin or erythrocin), CAS # 114-07-8, is a broad-spectrum antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Erythromycin is a macrolide antibiotic that works by binding the 50S ribosomal subunit and inhibiting polypeptide formation
  • Erythromycin was isolated from Saccharopolyspora erythraea

Applications

  • Erythromycin can be used to disrupt the growth of gram-positive and gram-negative bacteria
  • Erythromycin has been shown to have motor effects in the gastrointestinal system and can be used for laboratory research on migrating motor complex activity

Chemical Identifiers

CAS 114-07-8
Molecular Formula C37H67NO13
Molecular Weight (g/mol) 733.94
MDL Number MFCD00084654
InChI Key ULGZDMOVFRHVEP-RWJQBGPGSA-N
PubChem CID 12560
ChEBI CHEBI:42355
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Type (-)-Erythromycin
Physical Form Powder or Crystals
Melting Point 138.0°C to 140.0°C
Infrared Spectrum Authentic (Identification A)
Packaging Glass Bottle
Specific Rotation − 74.50 (20.00°C c=2,ethanol)
Water 6.5% max. (K.F.)
Color Colorless or White or Yellow
Quantity 5 g
Solubility Information Solubility in water: +/- 2 mg/mg. Other solubilities: freely soluble in alcohols, acetone, chloroform,, acetonitrile, ethyl acetate, moderately soluble in ether, ethylenedichloride, and amyl acetate
Formula Weight 733.94
Percent Purity 93.0 to 102.0% (sum of erythromycin A, B and C, on anhydrous substance); 5.0% max. erythromycin B and erythromycin C)
Chemical Name or Material (-)-Erythromycin
Show More Show Less
EINECSNumber 204-040-1
RTECSNumber KF4375000
TSCA TSCA

RUO – Research Use Only