missing translation for 'onlineSavingsMsg'
Learn More

Erucamide, ≥85% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012511
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012511 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012511 Supplier Merck Emd Millipore Supplier No. 9008225MG

Erucamide is a migratory additive that is commonly placed in polyolefin films to reduce their coefficient of friction (COF), a good attribute for high speed packaging operations.

Specifications

CAS 112-84-5
Linear Formula CH3(CH2)7 CH=CH(CH2)11 CONH2
MDL Number MFCD00882379
Melting Point 75°C to 85°C; 79°C to 81°C
Synonym cis-13-Docosenoamide; Erucamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 337.58
Percent Purity ≥85% (GC)
Grade Analytical Standard