missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Multispecies ERK1/ERK2 (Phospho) [pT202/pY204]/[pT185/pY187] InstantOne™ ELISA Kit

Catalog No. 8586012103
Encompass
Change view
Click to view available options
Quantity:
10 x 384 Tests
25 x 384 Tests
50 x 384 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8586012103 10 x 384 Tests
8586012253 25 x 384 Tests
8586012503 50 x 384 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 8586012103 Supplier Invitrogen™ Supplier No. 8586012103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 in cell lysates. Detection of rat ERK 1/2 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.

Specifications

Accession Number P27361,P28482
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5594, 5595
Gene Symbol ERK,ERK-2,ERK2,ERT1,MAPK2,P42MAPK,PRKM1,PRKM2,p38,p40,p41,p41mapk,p42-MAPK,ERK-1,ERK1,ERT2,HS44KDAP,HUMKER1A,P44ERK1,P44MAPK,PRKM3,p44-ERK1,p44-MAPK
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye HRP
Quantity 10 x 384 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 20 μL
Storage Requirements 2°C to 8°C
Target ERK1/2
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 10 mins
Show More Show Less