missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erioglaucine sodium salt
CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.844 g/mol
$47.36 - $108.11
Chemical Identifiers
| CAS | 3844-45-9 |
|---|---|
| Molecular Formula | C37H34N2Na2O9S3 |
| Molecular Weight (g/mol) | 792.844 |
| MDL Number | MFCD00012141 |
| InChI Key | SGHZXLIDFTYFHQ-UHFFFAOYSA-L |
| Synonym | C.I. 42090; Acid Blue 9 |
| PubChem CID | 19700 |
| ChEBI | CHEBI:82411 |
| IUPAC Name | disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate |
| SMILES | CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6178006
|
N/A | 5 g |
Each for $47.36
|
|
|||||
|
AAJ6178014
|
N/A | 25 g |
Each for $108.11
|
|
|||||
Description
Erioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsErioglaucine sodium salt is used as a colorant for foods and other substances to induce a color change.
Solubility
Soluble in water (30 mg/ml), ethanol.
Notes
Dye content: approx 60%
Chemical Identifiers
| 3844-45-9 | |
| 792.844 | |
| SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
| 19700 | |
| disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate |
| C37H34N2Na2O9S3 | |
| MFCD00012141 | |
| C.I. 42090; Acid Blue 9 | |
| CHEBI:82411 | |
| CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
| 283°C | |
| C37H34N2Na2O9S3 | |
| 14,1373 | |
| Soluble in water (30mg/ml),ethanol. | |
| CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] | |
| 792.844 | |
| CHEBI:82411 | |
| Purple | |
| Erioglaucine sodium salt |
| 3844-45-9 | |
| MFCD00012141 | |
| C.I. 42090; Acid Blue 9 | |
| SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
| disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate | |
| 19700 | |
| 792.86 | |
| Powder |
Safety and Handling
H500
EINECSNumber : 223-339-8
RTECSNumber : BQ4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only