Learn More
Thermo Scientific Chemicals Erioglaucine disodium salt, pure
CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.84 g/mol
Supplier: Thermo Scientific Chemicals 229730250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Erioglaucine disodium salt | |
| 1000 (at lambda min.) | |
| Purple | |
| Authentic | |
| C37H34N2Na2O9S3 | |
| 25 g | |
| 15, 1379 | |
| Solubility in water: soluble. Other solubilities: soluble in ethanol, insoluble in vegetable oils | |
| CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] | |
| 792.84 | |
| CHEBI:82411 | |
| Pure |
| 3844-45-9 | |
| 283.0°C | |
| 10% max. (105°C) | |
| Glass bottle | |
| MFCD00012141 | |
| 14, 855 | |
| brilliant blue fcf, acid blue 9, erioglaucine disodium salt, brilliant blue, FD&C blue no. 1, erioglaucine, alphazurine fg, FD&C blue no.1, fenazo blue xi, japan blue 1 | |
| SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
| disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate | |
| 19700 | |
| 792.84 | |
| Fine Crystalline Powder |
Chemical Identifiers
| 3844-45-9 | |
| 792.84 | |
| SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
| 19700 | |
| disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate |
| C37H34N2Na2O9S3 | |
| MFCD00012141 | |
| brilliant blue fcf, acid blue 9, erioglaucine disodium salt, brilliant blue, FD&C blue no. 1, erioglaucine, alphazurine fg, FD&C blue no.1, fenazo blue xi, japan blue 1 | |
| CHEBI:82411 | |
| CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 223-339-8
RTECSNumber : BQ4725000
TSCA : TSCA
RUO – Research Use Only