missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Erioglaucine disodium salt, pure

Catalog No. p-126330
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229730050 5 g Glass bottle
AC229730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229730050 Supplier Thermo Scientific Chemicals Supplier No. 229730050
Only null left

CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3844-45-9
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.84
MDL Number MFCD00012141
InChI Key SGHZXLIDFTYFHQ-UHFFFAOYSA-L
Synonym brilliant blue fcf, acid blue 9, erioglaucine disodium salt, brilliant blue, FD&C blue no. 1, erioglaucine, alphazurine fg, FD&C blue no.1, fenazo blue xi, japan blue 1
PubChem CID 19700
ChEBI CHEBI:82411
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Absorbance 1000 (at lambda min.)
Melting Point 283.0°C
Color Purple
Loss on Drying 10% max. (105°C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 14, 855
Merck Index 15, 1379
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, insoluble in vegetable oils
Formula Weight 792.84
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Erioglaucine disodium salt
Show More Show Less
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-339-8
RTECSNumber BQ4725000
TSCA TSCA

RUO – Research Use Only