missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eriochrome™ Black T, pure, indicator grade

Catalog No. ac228360250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228360250 25 g Glass bottle
AC228361000 100 g Glass bottle
AC228365000 500 g Glass bottle
AC228360010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC228360250 Supplier Thermo Scientific Chemicals Supplier No. 228360250
Only null left

CAS: 1787-61-7 | C20H12N3NaO7S | 461.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
Synonym C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11
PubChem CID 87355429
IUPAC Name (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Beilstein 16, III, 320
Merck Index 15, 3725
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
Formula Weight 461.38
Grade Pure
Packaging Glass bottle
Suitability Passes (for complexometric determination of metals)
UV Spectrometry Complies
Color Black to Black/Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eriochrome™ Black T
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-250-3
RTECSNumber QK2197000
TSCA TSCA

RUO – Research Use Only