missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome™ Black T, Dry Form in NaCl, for Hardness, Certified, LabChem™
$49.27 - $159.67
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Specifications
| 801°C | |
| 7647-14-5,1787-61-7 | |
| ClNa | |
| MFCD00003477 | |
| Passes Test | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Certified | |
| 2.165g/cm3 | |
| Hydrogen chloride; Carbon dioxide; Carbon monoxide; Nitrogen oxides; Sulfur compounds | |
| Purple | |
| Crystalline Powder |
| 1413°C | |
| 99.5,0.5 | |
| C20H12N3O7NaS | |
| Soluble in water | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 461.38 | |
| Poly Bottle | |
| Passes Test | |
| 2.165g/cm3 | |
| 100 g | |
| Eriochrome™ Black T, Dry Form in NaCl, for Hardness |
Safety and Handling
GHS H Statement
Mixture is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature