Learn More
Thermo Scientific Chemicals Eriochrome™ Black T, ACS reagent, indicator grade
CAS: 1787-61-7 | C20H12N3NaO7S | 461.38 g/mol
$144.25 - $336.58
Chemical Identifiers
| CAS | 1787-61-7 |
|---|---|
| Molecular Formula | C20H12N3NaO7S |
| Molecular Weight (g/mol) | 461.38 |
| MDL Number | MFCD00003935 |
| InChI Key | JHUJLRKQZAPSDP-GXTSIBQPSA-M |
| Synonym | C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11 |
| PubChem CID | 87355429 |
| SMILES | [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC439142500
|
Thermo Scientific Chemicals
439142500 |
250 g | Glass bottle |
Each for $144.25
|
|
||||
|
AC439140010
|
Thermo Scientific Chemicals
439140010 |
1 kg | Glass bottle |
Each for $336.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| C20H12N3NaO7S | |
| MFCD00003935 | |
| C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11 | |
| [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
Specifications
| 1787-61-7 | |
| C20H12N3NaO7S | |
| C.I. 14645, 3-Hydroxy-4-(1-hydroxy-2-naphthylazo)-7-nitro-1-naphthalene sulfonic acid, sodium salt, Mordant Black 11 | |
| JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
| (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium | |
| 87355429 | |
| ACS Reagent | |
| Black/Brown | |
| Eriochrome™ Black T |
| (0.1%, water) Passes Test | |
| MFCD00003935 | |
| Solubility in water: slightly soluble. | |
| [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O | |
| 461.38 | |
| 461.38 | |
| Suitability as complexometric indicator: Passes Test | |
| Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only