missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T 0.5% in Triethanolamine, Reagents
Specifications
Specifications
| Boiling Point | 335°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.1242 |
| Color | Purple |
| pH | 10.5 |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Percent Purity | 0.5% |
| Grade | Reagent |
| Chemical Name or Material | Eriochrome Black T 0.5% in Triethanolamine |