Learn More
Fisherbrand™ Erie Polyurethane Chair - High Bench Height with Round Tube Base and Stationary Glides in Polyurethane
Fisherbrand™ Erie Polyurethane Chairs with Round Tube Base are height adjustable for use at 42 in. work surfaces and feature handle in the back rest, fixed foot ring, and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$1,278.59 - $1,278.59
Specifications
| Adjustable | Yes |
|---|---|
| Back Style | Standard |
| With Armrest | No |
| With Backrest | Yes |
| With Lumbar Support | No |
Description
- Adjustable from 25.25-35 inches from the top of the seat to the floor
- Features manual back height and back depth adjustments
- Built in handle for ease of movement
- Black, powder-coated steel base with fixed foot ring
- Stationary, non-marring glides suitable for hard floors
- Chemical, soil, and puncture resistant polyurethane
Specifications
| Yes | |
| No | |
| No | |
| Polyurethane |
| Standard | |
| Yes | |
| No |