Learn More
Mettler Toledo™ ErgoSens Infrared Sensor for XPR Microbalances and Ultra-Microbalances
Description
- For touch-Less remote operation, USB connection
- For use with XPR Microbalances and Ultra-Microbalances
Specifications
Specifications
| For Use With (Equipment) | XPR Microbalance and Ultra-Microbalance |
| Connection Type | USB |
| For Use With (Application) | Touch-less remote operation |
| Type | Infrared Sensor |