Learn More
Ergodyne™ ProFlex™ 4000/4010 Wrist Supports
Flexible support that doesn't compromise productivity
Supplier:  Ergodyne™ 70022
 View more versions of this product
Catalog No. 19-035275C
Only null left
Explore available promotions
Description
Description
  • Ideal for material handling work
  • Durable supports encourage ergonomically neutral posture
  • Limit wrist flexion without compromising finger dexterity
  • Patented Open-Center Stay™ reduces pressure on median nerve
  • Fully adjustable for an exact fit and comfortable wear
  • Exclusive red line indicator ensures proper fit every time
  • Anti-odor treatment pulls moisture away from skin
  • Machine washable
ProFlex 4000
  • Limits downward wrist movement
  • ProFlex 4010
    • Adjustable double-closure stay that limits both upward and downward wrist movements
    Order Info
    Sold individually. Other styles available. See www.fishersafety.com.
    1 Measure around the smallest part of the wrist. This is a general guide only. Call your Fisher Safety Customer Service Representative at 1-800-772-6733 for more information.
    Specifications
    Specifications
    Black
    Proflex™ Single Strap Wrist Support
    Food processing, picking, packing, assembly, fabrication and taffy pulling
    Right
    Elastic
    6 in.
    Two stage hook and loop
    Small
    Patented open-center Stay™, red line indicator, anti-bacterial lining
    Limits both upward and downward wrist movements
    4010
    15.24 cm
    Product Suggestions
    Product Suggestions