missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERGIC3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ERGI3.
ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
Specifications
Specifications
| Antigen | ERGIC3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ERGIC3 |
| Gene Accession No. | Q9CQE7, Q9Y282 |
| Gene Alias | 2310015B14Rik; AV318804; C20orf47; CGI-54; D2Ucla1; dJ477O4.2; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; ERGIC and golgi 3; ERGIC3; ERV46; NY-BR-84; PRO0989; Sdbcag84; serologically defined breast cancer antigen 84; serologically defined breast cancer antigen NY-BR-84; Serologically defined breast cancer antigen NY-BR-84 homolog |
| Gene Symbols | ERGIC3 |
| Show More |