missing translation for 'onlineSavingsMsg'
Learn More

Erdosteine 98.0+%, TCI America™

Catalog No. E10261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10261G 1 g
E10265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10261G Supplier TCI America Supplier No. E10261G
Only null left

Chemical Identifiers

CAS 84611-23-4
Molecular Formula C8H11NO4S2
Molecular Weight (g/mol) 249.30
MDL Number MFCD00867664
InChI Key QGFORSXNKQLDNO-UHFFFAOYNA-N
Synonym N-(Tetrahydro-2-oxo-3-thienyl)-3-thiapentanedioic Acid Monoamide
PubChem CID 65632
IUPAC Name 2-({[(2-oxothiolan-3-yl)carbamoyl]methyl}sulfanyl)acetic acid
SMILES OC(=O)CSCC(=O)NC1CCSC1=O

Specifications

Melting Point 162°C
Color White
Quantity 1 g
Formula Weight 249.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Erdosteine
RTECSNumber AJ1476200
TSCA No