accessibility menu, dialog, popup

UserName

HOOSOHNSO

CAS RN 84611-23-4

IUPAC Name: 2-({[(2-oxothiolan-3-yl)carbamoyl]methyl}sulfanyl)acetic acid
Synonyms: erdosteine
Molecular Weight (g/mol): 249.3
Molecular Formula: C8H11NO4S2
InChi Key: QGFORSXNKQLDNO-UHFFFAOYNA-N
SMILES: OC(=O)CSCC(=O)NC1CCSC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results