missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Erbstatin Analog
An inhibitor of the EGF receptor associated tyrosine kinase
$156.23 - $275.60
Chemical Identifiers
| CAS | 63177-57-1 |
|---|---|
| Molecular Formula | C10H10O4 |
| Synonym | Methyl 2,5-dihydroxycinnamate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ61612LB0
|
Thermo Scientific Chemicals
J61612LB0 |
5 mg |
Each for $156.23
|
|
|||||
|
AAJ61612MA
|
Thermo Scientific Chemicals
J61612MA |
10 mg |
Each for $275.60
|
|
|||||
Description
An inhibitor of the EGF receptor associated tyrosine kinase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 63177-57-1 | |
| C10H10O4 | |
| Methyl 2,5-dihydroxycinnamate |
Specifications
| Yellow | |
| An inhibitor of the EGF receptor associated tyrosine kinase | |
| C10H10O4 | |
| Methyl 2,5-dihydroxycinnamate | |
| Signal Transduction Reagent | |
| BQCNSTFWSKOWMA-GORDUTHDSA-N | |
| methyl (2E)-3-(2,5-dihydroxyphenyl)prop-2-enoate |
| -30°C to -10°C | |
| 63177-57-1 | |
| 194.19g/mol | |
| Soluble in methanol or 0.1M NaOH; Insoluble in water | |
| Powder | |
| COC(=O)\C=C\C1=CC(O)=CC=C1O | |
| 194.19 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only