missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Erbstatin Analog

Catalog No. AAJ61612LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61612LB0 5 mg
AAJ61612MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61612LB0 Supplier Thermo Scientific Chemicals Supplier No. J61612LB0

An inhibitor of the EGF receptor associated tyrosine kinase

An inhibitor of the EGF receptor associated tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63177-57-1
Molecular Formula C10H10O4
Synonym Methyl 2,5-dihydroxycinnamate

Specifications

Quantity 5 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) An inhibitor of the EGF receptor associated tyrosine kinase
Formula Weight 194.19g/mol
Solubility Soluble in methanol or 0.1M NaOH; Insoluble in water
Product Type Signal Transduction Reagent
Form Powder
InChI Key BQCNSTFWSKOWMA-GORDUTHDSA-N
SMILES COC(=O)\C=C\C1=CC(O)=CC=C1O
IUPAC Name methyl (2E)-3-(2,5-dihydroxyphenyl)prop-2-enoate
Molecular Weight (g/mol) 194.19
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only