missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) perchlorate, 40 wt.% solution in water, Thermo Scientific™

Specifications

CAS 14017-55-1
Molecular Formula Cl3ErO12
Molecular Weight (g/mol) 465.61
MDL Number MFCD00016073
InChI Key FGBKFKCLWFRKEW-UHFFFAOYSA-K
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428290050
View Documents
Thermo Scientific Chemicals
428290050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC428290250
View Documents
Thermo Scientific Chemicals
428290250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent
Specifications

Specifications

14017-55-1
465.61
FGBKFKCLWFRKEW-UHFFFAOYSA-K
15335705
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]
Cl3ErO12
MFCD00016073
erbium 3+ perchlorate, erbium perchlorate, perchloric acid,erbium 3+ salt 8ci,9ci, erbium iii perchlorate solution, perchloric acid erbium iii salt, erbium perchlorate aqueous solution
erbium(3+);triperchlorate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only