accessibility menu, dialog, popup

UserName

CAS RN 14017-55-1

OClOOOOClOOOOClOOOEr

CAS RN 14017-55-1

IUPAC Name: erbium(3+) triperchlorate
Synonyms: erbium(3+) triperchlorate
Molecular Weight (g/mol): 465.6
Molecular Formula: Cl3ErO12
InChi Key: FGBKFKCLWFRKEW-UHFFFAOYSA-K
SMILES: [Er+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)