missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) oxide, REacton™, 99.9% (REO)

Catalog No. p-4412874
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1131014 25 g
AA1131022 100 g
AA11310A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1131014 Supplier Thermo Scientific Chemicals Supplier No. 01131014
Only null left

CAS: 12061-16-4 | Er2O3 | 382.52 g/mol

Erbium trioxide is an important rare earth metal used in biomedicine. It is used as a gate dielectric in semi conductor devices since it has a high dielectric constant and a large band gap; as a burnable neutron poison for nuclear fuel and as coloring for glasses. Erbium oxide nanoparticles can be surface modified for distribution into aqueous and non-aqueous media for bioimaging due to its photoluminescence property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12061-16-4
Molecular Formula Er2O3
Molecular Weight (g/mol) 382.52
MDL Number MFCD00010991
InChI Key ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
IUPAC Name dierbium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Er+3].[Er+3]

Specifications

Melting Point 2400°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 382.56
Sensitivity Hygroscopic
Density 8.64 g/mL
Chemical Name or Material Erbium(III) oxide
EINECSNumber 235-045-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only