Learn More
Erbium(III) oxalate decahydrate, 99.9%
CAS: 30618-31-6 | C6H26Er2O22 | 784.77 g/mol
$52.96 - $529.59
Chemical Identifiers
| CAS | 30618-31-6 |
|---|---|
| Molecular Formula | C6H26Er2O22 |
| Molecular Weight (g/mol) | 784.77 |
| MDL Number | MFCD00150142,MFCD00150142 |
| InChI Key | CYHKKMLSGVLNPD-UHFFFAOYSA-N |
| Synonym | erbium oxalate decahydrate, |
| PubChem CID | 131877482 |
| IUPAC Name | erbium;oxalic acid;decahydrate |
| SMILES | O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1842006
|
Thermo Scientific Chemicals
A1842006 |
5 g |
Each for $52.96
|
|
|||||
|
AAA1842014
|
Thermo Scientific Chemicals
A1842014 |
25 g |
Each for $159.93
|
|
|||||
|
AAA1842022
|
Thermo Scientific Chemicals
A1842022 |
100 g |
Each for $529.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 30618-31-6 | |
| 784.77 | |
| CYHKKMLSGVLNPD-UHFFFAOYSA-N | |
| 131877482 | |
| O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O |
| C6H26Er2O22 | |
| MFCD00150142,MFCD00150142 | |
| erbium oxalate decahydrate, | |
| erbium;oxalic acid;decahydrate |
Specifications
| 30618-31-6 | |
| C6H26Er2O22 | |
| MFCD00150142,MFCD00150142 | |
| UN3288 | |
| erbium oxalate decahydrate, | |
| O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O | |
| 784.77 | |
| 778.73 (598.58 Anhydrous) | |
| Erbium(III) oxalate decahydrate |
| 2.64 | |
| Er2(C2O4)3·10H2O | |
| 5 g | |
| Hygroscopic | |
| CYHKKMLSGVLNPD-UHFFFAOYSA-N | |
| erbium;oxalic acid;decahydrate | |
| 131877482 | |
| 99.9% |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only