Learn More
Erbium(III) oxalate decahydrate, 99.9%
CAS: 30618-31-6 | C6H26Er2O22 | 784.77 g/mol
Supplier: Thermo Scientific Chemicals A1842014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Erbium(III) oxalate decahydrate | |
| 2.64 | |
| Er2(C2O4)3·10H2O | |
| 25 g | |
| Hygroscopic | |
| CYHKKMLSGVLNPD-UHFFFAOYSA-N | |
| erbium;oxalic acid;decahydrate | |
| 131877482 | |
| 99.9% |
| 30618-31-6 | |
| C6H26Er2O22 | |
| MFCD00150142,MFCD00150142 | |
| UN3288 | |
| erbium oxalate decahydrate, | |
| O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O | |
| 784.77 | |
| 778.73 (598.58 Anhydrous) |
Chemical Identifiers
| 30618-31-6 | |
| 784.77 | |
| CYHKKMLSGVLNPD-UHFFFAOYSA-N | |
| 131877482 | |
| O.O.O.O.O.O.O.O.O.O.[Er].[Er].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O |
| C6H26Er2O22 | |
| MFCD00150142,MFCD00150142 | |
| erbium oxalate decahydrate, | |
| erbium;oxalic acid;decahydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only