Learn More
ErbB2 (HER-2) Monoclonal Antibody (MJD2), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$722.00 - $722.00
Specifications
| Antigen | ErbB2 (HER-2) |
|---|---|
| Clone | MJD2 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Description: This MJD2 monoclonal antibody reacts with HER-2, the product of the ErbB2 gene and a member of the human epidermal growth factor receptor (EGFR) family. HER-2 is a 185 kDa transmembrane receptor with an extracellular binding domain and an intracellular tyrosine kinase domain. Activation of HER-2 occurs following formation of homodimers or heterodimers with ligand-bound EGFR family members, HER3 or HER4. Overexpression of HER-2 results in auto-phosphorylation and increased proliferation and invasiveness and is associated with subsets of multiple cancer types such as breast, ovarian, cervical, uterine, and gastric. The use of HER-2 targeted agents has been used extensively to treat HER-2-positive breast cancer, but a recent study found that these agents can also block the proliferation of breast cancer stem cells found in HER-2-negative patients and suggests the importance of HER-2 as a target for multiple types of breast cancer. The MJD2 antibody recognizes the extracellular domain of HER-2 but at an epitope distinct from that recognized by the 4D5 (Herceptin) and SP3 monoclonal antibodies. Applications Reported: This MJD2 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.Specifications
| ErbB2 (HER-2) | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| 2064 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MJD2 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P04626 | |
| ERBB2 | |
| Primary | |
| 4°C | |
| ERBB2 |
For Research Use Only.