missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Equol 98.0+%, TCI America™

Catalog No. E0922200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0922200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0922200MG Supplier TCI America Supplier No. E0922200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94105-90-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00016662
InChI Key ADFCQWZHKCXPAJ-UHFFFAOYNA-N
Synonym 3-4-hydroxyphenyl chroman-7-ol, equol, +/--equol, r,s-equol, 7,4'-homoisoflavane, 4',7-dihydroxyisoflavane, rs-equol, 3-4-hydroxyphenyl-7-chromanol, 2h-1-benzopyran-7-ol, 3,4-dihydro-3-4-hydroxyphenyl, 4',7-isoflavandiol
PubChem CID 382975
IUPAC Name 3-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol
SMILES OC1=CC=C(C=C1)C1COC2=C(C1)C=CC(O)=C2

Specifications

Melting Point 175°C
Quantity 200 mg
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Equol