missing translation for 'onlineSavingsMsg'
Learn More

Hughes Equipment Inspection Record

Catalog No. 18666723
Change view
Click to view available options
Equipment Inspection Record
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18666723 Equipment Inspection Record
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18666723 Supplier Justrite International Supplier No. SERVICECARDS

Equipment Inspection Record can be attached to the shower with the Equipment Inspection Record Holder or positioned close by where it can be readily available.

Regular servicing and testing of Emergency Safety Showers and Eye/Facewash units is critical to ensuring proper functionality in the event of an emergency. Depending on the location and company procedures you may test daily or weekly and carry out servicing every 3, 6 or 12 months. Each time the equipment is tested or serviced in accordance with the agreed schedule, the person responsible dates and signs the tag. When the tag is full, simply remove it from the holder and replace with a new inspection record. As a best practice, completed tags should be kept as a record of testing and servicing.

Features:

  • Document comprehensive service/test history for the life of the unit
  • Helps make identifying the site location and model simple
  • Plastic material ensures written records will last, even when exposed to water
  • Includes two blank tags with space for 26 service records

Compliance

Not ANSI Compliant

Warranty

1 Year

Specifications

Color White, Black, Red
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 6.5 in.
Height (Metric) 165 mm
Includes 2 Pk.
Material Plastic
Product Type Safety Shower and Emergency Eye Wash
Width (English) 2 in.
Width (Metric) 51 mm