missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EquiPhi29™ DNA Amplification Kit

Catalog No. p-200009440
Change view
Click to view available options
Quantity:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65393 100 Reactions
A65394 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A65393 Supplier Thermo Scientific™ Supplier No. A65393
Only null left

The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).

The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).

Features of the EquiPhi29 DNA Amplification Kit include:
• Fast DNA amplification in less than two hours
• Sensitivity from 1 fg of input DNA
• Maximal yield of 17 µg amplified DNA
• Compatibility with different types of sample input material (e.g., purified DNA, liquid media culture, agar plate colonies)
• Compatibility with various downstream applications such as DNA sequencing (Sanger sequencing, next-generation sequencing), digestion with restriction enzymes, cell-free DNA enrichment, and cell-free protein expression
• All required components for DNA amplification in RCA or MDA-WGA reactions

The kit contains EquPhi29 DNA Polymerase, a proprietary phi29 DNA polymerase mutant that possesses strong strand displacement activity and enables high thermostability, reaction speed, and product yield and low amplification bias. Also included is an optimized reaction buffer, exo-resistant random primer, pyrophosphatase, and dNTPs. The addition of pyrophosphatase to the reaction mixture enhances DNA amplification and facilitates sample handling.

Specifications

Product Type DNA Amplification Kit
Form Liquid
For Use With (Application) RCA, MDA-WGA
Polymerase EquiPhi29 DNA Polymerase
Quantity 100 Reactions
Research Category Nucleic Acid Analysis, In Vitro Transcription, Cell-free Protein Expression, In Situ Sequencing, SNP Detection
Content And Storage

• EquiPhi29 DNA Polymerase (0.1 mL at 10 U/μL)
• 10X EquiPhi29 DNA Polymerase Reaction Buffer (1 mL)
• 0.1 M DTT (0.25 mL)
• 500 μM Exo-Resistant Random Primer (0.2 mL)
• 10 mM dNTP Mix (0.2 mL)
• 0.1 U/μL Pyrophosphatase, inorganic (0.1 mL)

Store at -15 to -25°C.

Format Kit
Optimal Reaction Temperature 42°C
Shipping Condition Dry Ice
Reaction Time Up to 2 hours
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.