missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Equine TNF alpha Uncoated ELISA Kit

Catalog No. ENESS0017
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENESS0017 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENESS0017 Supplier Invitrogen™ Supplier No. ESS0017
Only null left

ELISA

The Equine TNF-alpha Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of equine TNFalpha. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.

Specifications

Accession Number P29553
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity <15.6 pg/mL
Description Equine TNF alpha ELISA Reagent Kit
Conjugate HRP
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA
Sample Type Serum, Supernatant
Specificity No significant cross-reaction with bovine, mouse, or rat TNFa. Partial cross-reaction occurs with human and porcine TNFa (<10%)
Sufficient For 5 x 96 microplate wells
Target Species Equine
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 100033834
Gene Symbol TNF-a,TNF-alpha,TNFA,TNFSF2
Kit Contents Coating Antibody, Lyophilized Recombinant Standard, Detection Antibody, Streptavidin-HRP, Substrate Solution, Ready to use Stop Solution, 0.16M Sulfuric Acid, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
Label or Dye HRP
Measurement Absorbance at 450 nm minus absorbance (at 550 nm)
Precision Intra-assay CV <10%; inter assay CV <10%
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 100 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Target TNF alpha
Test Time 24 hrs
Total Assay Time 2 hrs 50 mins
Show More Show Less