missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epstein Barr Virus p18 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA173003
Description
The PA1-73003 antibody reacts with Epstein-Barr virus p18. PA1-73003 has been successfully used in ELISA and immunofluorescence applications. IHC and WB are potential applications but have not yet been tested. The PA1-73003 antibody was raised against Recombinant EBV p18. Recommended short term storage (<6 months) is at 4°C. For longer term storage, aliquot and freeze.
Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.Chemical Identifiers
| Epstein Barr Virus p18 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Virus | |
| EBV p18 | |
| Primary | |
| IgG fraction |
| 5.0 mg/mL | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Goat | |
| RUO | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Recombinant EBV p18. | |
| Antibody |
Specifications
| Epstein Barr Virus p18 | |
| Polyclonal | |
| Unconjugated | |
| EBV p18 | |
| Recombinant EBV p18. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 5.0 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |