missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactam 99.0+%, TCI America™

Catalog No. C0203500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0203500G 500 g
C020325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0203500G Supplier TCI America Supplier No. C0203500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-60-2
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006936
InChI Key JBKVHLHDHHXQEQ-UHFFFAOYSA-N
Synonym epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
PubChem CID 7768
ChEBI CHEBI:28579
IUPAC Name azepan-2-one
SMILES C1CCC(=O)NCC1

Specifications

Melting Point 70°C
Color White
Boiling Point 267°C
Quantity 500 g
Formula Weight 113.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material epsilon-Caprolactam
EINECSNumber (5)-1097
RTECSNumber CM3675000
TSCA Yes