missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EPPS, 99%, for biochemistry

$230.66 - $624.27

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym 3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC172601000
View Documents Promotion Details
Thermo Scientific™
172601000
100g Glass bottle
Each for $230.66
Only null left
 
AC172605000
View Documents Promotion Details
Thermo Scientific™
172605000
500g Glass bottle
Each for $624.27
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

(0.1 M aq. soln.) (1 cm cell)
16052-06-5
99%
C9H20N2O4S
3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
≥98.5%
EPPS
(0.1M aq. soln.) (1 cm cell)
5 to 6 (10% w/v soln. at 20°C)
Authentic
MFCD00006160
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
252.33
Biochemical
Videos
SDS
Documents

Documents

RUO – Research Use Only