missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EPPS, 99%, for biochemistry

Supplier:  Thermo Scientific™ 172601000

 View more versions of this product

Catalog No. AC172601000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Specifications

(0.1 M aq. soln.) (1 cm cell)
(0.1M aq. soln.) (1 cm cell)
5 to 6 (10% w/v soln. at 20°C)
99%
C9H20N2O4S
Glass bottle
OWXMKDGYPWMGEB-UHFFFAOYSA-N
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
85255
252.33
Biochemical
EPPS
16052-06-5
100g
Authentic
MFCD00006160
3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
C1CN(CCN1CCCS(=O)(=O)O)CCO
252.329
CHEBI:42298
≥98.5%

Chemical Identifiers

16052-06-5
252.329
OWXMKDGYPWMGEB-UHFFFAOYSA-N
85255
3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
C9H20N2O4S
MFCD00006160
3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
CHEBI:42298
C1CN(CCN1CCCS(=O)(=O)O)CCO
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only