Learn More
Eppendorf™ Trade-in Xplorer™ Electronic Pipetters, Multichannel
Specially designed for high professional standards to provide optimal support.
Supplier: Eppendorf™ 486100156
Description
Pipetters are based on the proven Eppendorf PhysioCare™ concept. The features set new standards for simplicity, precision and reproducibility, which mean no more delays due to complicated programming or inflexible processes. Precisely adjustable parameters, maximally reproducible results, fatigue-free work and full control over the pipetting processes are achieved.
- Intuitive user interface
- Pipetting modes: autodispensing, dispensing, pipetting, mixing, manual pipetting
- Universal nose cone fits most pipette tips
- Highly reproducible results
- Spring engaged nose cone for enhanced tip attachment
- Light weight design offers unsurpassed ergonomics
Chemical Identifiers
| Trade-in Xplorer | |
| Electronic Pipette | |
| Ergonomic |
Specifications
| Orange | |
| Trade-in Xplorer | |
| 15 to 300 μL |
| 12 | |
| Electronic Pipette | |
| Ergonomic |