Learn More
Eppendorf™ Trade-in Xplorer™ Electronic Pipetters, Multichannel
Specially designed for high professional standards to provide optimal support.
Supplier:  Eppendorf™ 486100121
 View more versions of this product
Catalog No. 14286539
Only null left
Description
Description
Pipetters are based on the proven Eppendorf PhysioCare™ concept. The features set new standards for simplicity, precision and reproducibility, which mean no more delays due to complicated programming or inflexible processes. Precisely adjustable parameters, maximally reproducible results, fatigue-free work and full control over the pipetting processes are achieved.
  • Intuitive user interface
  • Pipetting modes: autodispensing, dispensing, pipetting, mixing, manual pipetting
  • Universal nose cone fits most pipette tips
  • Highly reproducible results
  • Spring engaged nose cone for enhanced tip attachment
  • Light weight design offers unsurpassed ergonomics
Specifications
Chemical Identifiers
Trade-in Xplorer
Electronic Pipette
Ergonomic
Specifications
Yellow
Trade-in Xplorer
5 to 100 μL
8
Electronic Pipette
Ergonomic
Product Suggestions
Product Suggestions