missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube

Catalog No. p-194466
Change view
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Biopur™
Eppendorf Quality
PCR Clean
Max. RCF:
22,000 x g
30,000 x g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade Max. RCF
0540227 Blue Eppendorf Quality 30,000 x g
0540228 Green Eppendorf Quality 30,000 x g
0540229 Red Eppendorf Quality 30,000 x g
0540230 Yellow Eppendorf Quality 30,000 x g
0540225 Clear Eppendorf Quality 30,000 x g
054025 Assorted Eppendorf Quality 30,000 x g
0540294 Clear PCR Clean 30,000 x g
0540226 Amber PCR Clean 30,000 x g
0540224B Clear Biopur™ 22,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 0540227 Supplier Eppendorf™ Supplier No. 022363247
Only null left

Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube ensures secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 mL capacity.

  • The hinged Eppendorf Safe-Lock lid prevents inadvertent lid opening during incubation and storage.
  • g-Safe™ technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30,000 x g, preventing sample loss due to tube breakage and providing additional safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances demonstrated to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precision lid sealing minimizes evaporation rates during extended storage periods.
  • The generously sized frosted lid and side surface facilitate easy labeling.
  • Assured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121 °C, 20 min) for reliable sterilization.
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.

Order Info

Supplied individually wrapped in your choice of three sizes

Specifications

Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Blue
Material Polypropylene
No. per Pack 500 Pk.
Packaging 500 Tubes (1 Bag x 500 Tubes)
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Clean Claims Free of plasticizers, biocides or mold release agents
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.05 oz.
Closure Type Safe-lock Lid
Length (English) 1.5 in.
Bottom Shape Conical
Max. RCF 30,000 x g
Diameter (Metric) 10 mm
Shape Cylindrical
Length (Metric) 38 mm
Show More Show Less