Learn More
Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rectangular Bucket Adapters
Bucket adapters for use with Eppendorf 5804R Centrifuge.
Supplier:  Eppendorf™ 022637681
 View more versions of this product
Catalog No. 36-100-7193
Only null left
Explore available promotions
Description
Description
For use with Standard and Vacutainer conical tubes and micro test tubes.
Specifications
Specifications
Bucket Adapter
31 mm
30 to 50 mL
Rectangular Bucket (100 mL)
1
Standard and Vacutainer conical tubes and micro test tubes
Product Suggestions
Product Suggestions